Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclohexylmethane 4,4'-diisocyanate, mixture of isomers, tech. 90%

Chemical Identifiers

Chemical Name or Material Dicyclohexylmethane 4,4'-diisocyanate
CAS 5124-30-1
Name Note Mixture of isomers
Purity Grade Technical Grade
Molecular Formula C15H22N2O2
Formula Weight 262.37
MDL Number MFCD00015405
UN Number UN2206
Synonym 4,4'-Methylenebis(cyclohexyl isocyanate)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44087AD

  SDS
alfa aesar™
44087AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44087AK

  SDS
alfa aesar™
44087AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 262.37
Chemical Name or Material Dicyclohexylmethane 4,4'-diisocyanate
Physical Form Liquid
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Health Hazard 1 Danger
RTECS Number NQ9250000
Quantity 50mL
MDL Number MFCD00015405
Synonym 4,4'-Methylenebis(cyclohexyl isocyanate)
Health Hazard 2 GHS H Statement
H331-H334-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Boiling Point 168°C (1.5mmHg)
Density 1.066g/mL
TSCA TSCA
CAS 5124-30-1
Name Note Mixture of isomers
Molecular Formula C15H22N2O2
Purity Grade Technical Grade
Assay 90%
UN Number UN2206
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.