missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Dicyclohexylamine, ≥99.5% (GC), Honeywell™ Fluka™

for titrimetric determination of isocyanates,≥99.5% (GC)

$71.60 - $273.09

Chemical Identifiers

CAS 101-83-7
Molecular Formula C12H23N
Molecular Weight (g/mol) 181.323
MDL Number MFCD00011658
InChI Key XBPCUCUWBYBCDP-UHFFFAOYSA-N
Synonym dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
PubChem CID 7582
ChEBI CHEBI:34694
IUPAC Name N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC2CCCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046645 View Documents Honeywell Fluka™
36620100ML
100mL Glass Bottle Each for $71.60
 
60046646 View Documents Honeywell Fluka™
36620500ML
500mL Glass Bottle Each for $273.09
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

101-83-7
181.323
XBPCUCUWBYBCDP-UHFFFAOYSA-N
7582
N-cyclohexylcyclohexanamine
C12H23N
MFCD00011658
dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
CHEBI:34694
C1CCC(CC1)NC2CCCCC2

Specifications

Liquid
96°C (204.8°F)
n20/D 1.484
12mmHg (37.7°C)
Colorless to Yellow
100mL
C12H23N
MFCD00011658
605923
XBPCUCUWBYBCDP-UHFFFAOYSA-N
N-cyclohexylcyclohexanamine
7582
181.32g/mol
Dicyclohexylamine
0.912g/mL at 20°C
Glass Bottle
6 (vs air)
117°C to 120°C
-2°C
101-83-7
(C6H11)2NH
UN2565
dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
C1CCC(CC1)NC2CCCCC2
181.323
CHEBI:34694
≥99.5% (GC)
Safety and Handling

Safety and Handling

P273-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

DOTInformation : 8/PGIII

RTECSNumber : 202-980-7