missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dicyclohexyl phthalate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5600436
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dicyclohexyl phthalate | |
63°C to 67°C | |
C20H26O4 | |
MFCD00003849 | |
1889288 | |
VOWAEIGWURALJQ-UHFFFAOYSA-N | |
dicyclohexyl benzene-1,2-dicarboxylate | |
6777 | |
330.42 |
84-61-7 | |
1.383 | |
1.431 | |
500 g | |
dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 | |
330.424 | |
CHEBI:34693 | |
≥99% |
Chemical Identifiers
84-61-7 | |
330.424 | |
VOWAEIGWURALJQ-UHFFFAOYSA-N | |
6777 | |
dicyclohexyl benzene-1,2-dicarboxylate |
C20H26O4 | |
MFCD00003849 | |
dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
CHEBI:34693 | |
C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H360D
EINECSNumber : 201-545-9
RTECSNumber : TI0889000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only