missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclohexyl phthalate, 99+%

Manufacturer:  Alfa Aesar™ H5600422

 View more versions of this product

Catalog No. AAH5600422



Specifications

Specifications

Dicyclohexyl phthalate
100g
C20H26O4
1889288
VOWAEIGWURALJQ-UHFFFAOYSA-N
dicyclohexyl benzene-1,2-dicarboxylate
6777
330.42
63°C to 67°C
84-61-7
MFCD00003849
1,2-benzenedicarboxylic acid, dicyclohexyl ester
C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3
330.424
CHEBI:34693
≥99%

Chemical Identifiers

84-61-7
330.424
VOWAEIGWURALJQ-UHFFFAOYSA-N
6777
dicyclohexyl benzene-1,2-dicarboxylate
C20H26O4
MFCD00003849
1,2-benzenedicarboxylic acid, dicyclohexyl ester
CHEBI:34693
C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TI0889000

TSCA : TSCA

Documents