Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclohexyl disulfide, 98%

$88.59 - $277.24

Chemical Identifiers

Chemical Name or Material Dicyclohexyl disulfide
CAS 2550-40-5
Molecular Formula C12H22S2
Formula Weight 230.44
MDL Number MFCD00013759
Beilstein 1905920
Synonym Cyclohexyl disulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1218214

  SDS
alfa aesar™
L1218214
25g Each for $88.59

AAL1218222

  SDS
alfa aesar™
L1218222
100g Each for $277.24
Description & Specifications

Specifications

Formula Weight 230.44
Chemical Name or Material Dicyclohexyl disulfide
Assay 98%
Molecular Formula C12H22S2
MDL Number MFCD00013759
CAS 2550-40-5
Beilstein 1905920
Synonym Cyclohexyl disulfide
RTECS Number JO1843850
TSCA TSCA
Odor Unpleasant
Boiling Point 331°C
Flash Point 170°C (338°F)
Quantity 25g

Applications
Dicyclohexyl disulfide is employed as an edible spices. It is also used in dyes, medicine and organic synthesis. It is also useful as a synthetic flavor ingredient. Further, it is used in the production of n-(cyclohexylthio) phthalimide.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.