missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine, 97%

Manufacturer:  Alfa Aesar™ H6191406

 View more versions of this product

Catalog No. AAH6191406



Specifications

Specifications

Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine
138°C to 140°C
303111-96-8
MFCD11656073
DTSPXGRQYHLKLO-UHFFFAOYSA-N
dicyclohexyl-[2,4,6-tri(propan-2-yl)phenyl]phosphane
9801140
Solid
Air Sensitive
5g
C27H45P
dicyclohexyl 2,4,6-triisopropylphenyl phosphine, 2,4,6 tri-isopropyl phenyl di-cyclohexylphosphine, 2,4,6-tri-isopropyl phenyl di-cyclohexylphosphine, dicyclohexyl 2,4,6-tris 1-methylethyl phenyl phosphine, dicyclohexyl 2,4,6-triisopropylphenyl phosphane, acmc-1ajbl, 2,4,6-tri-isopropyl phenyl di-cyclohexylphosphine, 2,4,6 triisopropyl phenyl dicyclohexylphosphine, 1,3,5-triisopropyl-2-dicyclohexylphosphino benzene, dicyclohexyl 2,4,6-tri propan-2-yl phenyl phosphane
CC(C)C1=CC(=C(C(=C1)C(C)C)P(C2CCCCC2)C3CCCCC3)C(C)C
400.631
400.62
97%

Chemical Identifiers

303111-96-8
400.631
DTSPXGRQYHLKLO-UHFFFAOYSA-N
9801140
CC(C)C1=CC(=C(C(=C1)C(C)C)P(C2CCCCC2)C3CCCCC3)C(C)C
C27H45P
MFCD11656073
dicyclohexyl 2,4,6-triisopropylphenyl phosphine, 2,4,6 tri-isopropyl phenyl di-cyclohexylphosphine, 2,4,6-tri-isopropyl phenyl di-cyclohexylphosphine, dicyclohexyl 2,4,6-tris 1-methylethyl phenyl phosphine, dicyclohexyl 2,4,6-triisopropylphenyl phosphane, acmc-1ajbl, 2,4,6-tri-isopropyl phenyl di-cyclohexylphosphine, 2,4,6 triisopropyl phenyl dicyclohexylphosphine, 1,3,5-triisopropyl-2-dicyclohexylphosphino benzene, dicyclohexyl 2,4,6-tri propan-2-yl phenyl phosphane
dicyclohexyl-[2,4,6-tri(propan-2-yl)phenyl]phosphane
Documents