Please login to your online account to display your discounted pricing

Dicyanobis(1,10-phenanthroline)iron(II) dihydrate, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391320010

 View more versions of this product

Catalog No. AC391320010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 504.33
Physical Form Powder
Quantity 1g
Color Dark brown
Suitability Passes Test
Chemical Name or Material Dicyanobis(1, 10-phenanthroline)iron(II) dihydrate
CAS 23425-29-8
Molecular Formula C26H16FeN6·2H2O
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00150644
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Contact with acids liberates toxic gas.
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Dicyanobis(1, 10-phenanthroline)iron(II) dihydrate
CAS 23425-29-8
Molecular Formula C26H16FeN6·2H2O
Formula Weight 504.33
MDL Number MFCD00150644
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Contact with acids liberates toxic gas.

SDS & Safety Documents