missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dicyanine A 98%, ACROS Organics™

Chemical Identifiers

CAS 20591-23-5
Assay Percent Range 98%
Molecular Formula C31H37IN2O2
Formula Weight 596.54
MDL Number MFCD00051379
Synonym 6-Ethoxy-2-[3-(6-ethoxy-1-ethyl-2-methyl-4-quinolylidene)]propenyl-1-ethyl-4-methyl-quinolinium iodide
Chemical Name or Material Dicyanine A, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC407050010 SDS Fisher Scientific Acros
407050010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

20591-23-5
C31H37IN2O2
MFCD00051379
Dicyanine A, 98%
98%
596.54
6-Ethoxy-2-[3-(6-ethoxy-1-ethyl-2-methyl-4-quinolylidene)]propenyl-1-ethyl-4-methyl-quinolinium iodide

Specifications

1g
Authentic
Crystalline Powder
98%
596.54
Green
6-Ethoxy-2-[3-(6-ethoxy-1-ethyl-2-methyl-4-quinolylidene)]propenyl-1-ethyl-4-methyl-quinolinium iodide
20591-23-5
Glass bottle
97.5% min.
C31H37IN2O2
MFCD00051379
23, IV, 2069
Dicyanine A, 98%

Safety and Handling

EINECSNumber : 243-897-6

TSCA : TSCA