missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dicyandiamide, 99.5%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC113810250
Specifications
Dicyandiamide | |
Plastic bottle | |
0.5% max. (K.F.) | |
208.0°C to 211.0°C | |
461-58-5 | |
C2H4N4 | |
01,229 | |
1-cyanoguanidine, cyanoguanidine, dicyandiamide, dicyandiamido, dicyandiamin, dicyanodiamide, epicure dicy 7, guanidine, cyano, n-cyanoguanidine, pyroset do | |
QGBSISYHAICWAH-UHFFFAOYSA-N | |
2-cyanoguanidine | |
10005 | |
Crystalline Powder |
Authentic | |
0.1% max. | |
White | |
25g | |
99.5% | |
MFCD00008066 | |
15, 3103 | |
Solubility in water: 32g/L (20°C). Other solubilities: 38g/L in methanol (20°C) | |
C(#N)N=C(N)N | |
84.08 | |
84.08 | |
99.5% |
Chemical Identifiers
461-58-5 | |
84.08 | |
QGBSISYHAICWAH-UHFFFAOYSA-N | |
10005 | |
C(#N)N=C(N)N |
C2H4N4 | |
MFCD00008066 | |
1-cyanoguanidine, cyanoguanidine, dicyandiamide, dicyandiamido, dicyandiamin, dicyanodiamide, epicure dicy 7, guanidine, cyano, n-cyanoguanidine, pyroset do | |
2-cyanoguanidine |
Safety and Handling
EINECSNumber : 207-312-8