missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dicyandiamide, 99.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 113810010

 View more versions of this product

Catalog No. AC113810010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dicyandiamide
208.0°C to 211.0°C
Authentic
Plastic bottle
MFCD00008066
01,229
dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
QGBSISYHAICWAH-UHFFFAOYSA-N
N''-cyanoguanidine
10005
99.5%
461-58-5
White
99.5%
C2H4N4
1 kg
15, 3103
Solubility in water: 32g/L (20°C). Other solubilities: 38g/L in methanol (20°C)
NC(N)=NC#N
84.08
84.08
Crystalline Powder

Chemical Identifiers

461-58-5
84.08
QGBSISYHAICWAH-UHFFFAOYSA-N
10005
C2H4N4
MFCD00008066
dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
NC(N)=NC#N
Safety and Handling

Safety and Handling

EINECSNumber : 207-312-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only