Learn More
Dicumarol, Thermo Scientific Chemicals
Vitamin K reductase inhibitor possessing anticoagulant properties
Supplier: Thermo Scientific Chemicals J6363414
Description
Dicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.
Solubility
Soluble in aqueous alkaline solutions, organic bases, 0.1 N NaOH (15 mg/ml), Pyridine (50 mg/ml), chloroform (slightly soluble), and benzene (slightly soluble). Insoluble in water, and alcohols.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Dicumarol | |
286°C to 289°C | |
C19H12O6 | |
25 g | |
14,3090 | |
Soluble in aqueous alkaline solutions,organic bases,0.1 N NaOH (15mg/ml),Pyridine (50mg/ml),chloroform (slightly soluble),and benzene (slightly soluble). Insoluble in water,and alcohols. | |
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12 | |
336.30 | |
CHEBI:4513 | |
Powder |
66-76-2 | |
White | |
MFCD00006857 | |
UN2811 | |
dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl | |
DOBMPNYZJYQDGZ-UHFFFAOYSA-N | |
4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one | |
54676038 | |
336.3 |
Chemical Identifiers
66-76-2 | |
336.30 | |
DOBMPNYZJYQDGZ-UHFFFAOYSA-N | |
54676038 | |
4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one |
C19H12O6 | |
MFCD00006857 | |
dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl | |
CHEBI:4513 | |
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12 |
Safety and Handling
GHS H Statement
H372-H302
Causes damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P260-P264b-P270-P301+P310-P314-P330-P501c
H301-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-632-9
RTECSNumber : GN7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only