missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dicobalt octacarbonyl, 95%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC291841000
Specifications
Dicobalt Octacarbonyl | |
−13°C | |
Glass Bottle | |
5% max. hexane | |
51.0°C to 52.0°C | |
95% | |
94% min | |
Co2(CO)8 | |
3124 | |
carbon monoxide; cobalt 2+, cobalt carbonyl, moistened with hexane hexane co, dicobalt octacarbonyl stabilised with hexane | |
341.95 | |
Crystals or Flakes |
1.8100g/mL | |
Authentic | |
1.81 | |
Orange-Red to Red | |
100g | |
10210-68-1 | |
C8Co2O8 | |
MFCD00016024 | |
15,3095 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and carbon disulfide | |
341.95 | |
95% |
Chemical Identifiers
10210-68-1 | |
341.95 | |
carbon monoxide; cobalt 2+, cobalt carbonyl, moistened with hexane hexane co, dicobalt octacarbonyl stabilised with hexane |
C8Co2O8 | |
MFCD00016024 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Self-heating; may catch fire.
GHS P Statement
Keep cool.
Protect from sunlight.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty
GHS Signal Word: Danger
EINECSNumber : 233-514-0
RTECSNumber : GG0300000
TSCA : TSCA