Dickson™ Waterproof Temperature Data Logger
Food-grade stainless-steel, waterproof datalogger
Manufacturer:  Dickson™ HT200
 View more versions of this product
Catalog No. 15-075-132
Specifications
Specifications
Tadiran battery and quick start guide
±1.8°F
1 Year
0.75 in.
68
For HACCP, FDA, pharmaceutical, medical agricultural and oil and gas applications
13.6cm
5.21 oz.
HACCP, FDA
Tadiran TLH-5955 Lithium
Temperature
1.9cm
Temperature Datalogger
5.36 in.
-40° to 125°C
148g
Documents