missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dickson™ Replacement Charts for Super-Compact SC3 Temperature Chart Recorders

For SC3 Temperature Recorders.

Manufacturer:  Dickson™ C306

 View more versions of this product

Catalog No. 15-074-67



Description

Description

  • Seven-day, 3 in. diameter (76mm) charts
  • Comes in package of 60 sheets
Specifications

Specifications

3 in.
7.6cm
For super-compact SC3 temperature chart
Fahrenheit chart
50° to 96°F
10° to 35°C
Documents