missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dickson™ Pro Series USB Temperature Datalogger

Temperature data logger excels in the widest range of applications, regulatory compliance, HVAC balancing, warehouse mapping, process control and more

Manufacturer:  Dickson™ SP125

 View more versions of this product

Catalog No. 15-174-110



Description

Description

  • USB enabled fast downloading
  • 32KB data storage
  • User defined sampling rates - you determine how often it takes a reading
  • Battery powered

Specifications

Specifications

-23° to 80°C
CE
±1.2°F/±0.7°C
1
20
For widest range of applications, regulatory compliance, HVAC balancing, warehouse mapping, process control and more
53g
CR2450 Lithium battery and quick start guide
LED Indicator, Visual
1 Year
32512
Temperature Datalogger
19 oz.
Documents