Please login to your online account to display your discounted pricing
Dickson™ Pro Series USB Temperature Datalogger
Temperature data logger excels in the widest range of applications, regulatory compliance, HVAC balancing, warehouse mapping, process control and more
$250.96 - $482.96
Specifications
Alarm Types LED Indicator, Visual
Battery Life 1 Year
Certifications/Compliance CE
Product Type Temperature Datalogger
For Use With (Application) For widest range of applications, regulatory compliance, HVAC balancing, warehouse mapping, process control and more
View More Specs
Products
Catalog Number Mfr. No. Accuracy (Temperature) Channels Data Memory Temperature Range (Metric) Price Quantity    
15-174-111
dickson™
SP175
±1.8°F/±1°C 2 16256 -150° to 1093°C Each for $482.96
15-174-110
dickson™
SP125
±1.2°F/±0.7°C 1 32512 -23° to 80°C Each for $250.96
Description & Specifications
Specifications
Alarm Types LED Indicator, Visual
Battery Life 1 Year
Certifications/Compliance CE
Product Type Temperature Datalogger
For Use With (Application) For widest range of applications, regulatory compliance, HVAC balancing, warehouse mapping, process control and more
IP Rating 20
Weight (English) 19 oz.
Weight (Metric) 53g
  • USB enabled fast downloading
  • 32KB data storage
  • User defined sampling rates - you determine how often it takes a reading
  • Battery powered