Dickson™ Accessories for LCD Display Temperature Data Loggers
Supports easy data collection
Manufacturer:  Dickson™ A220
 View more versions of this product
Catalog No. 15-075-116
Includes: Software: USB downloadable cable; Card reader kit: card reader and removable SD flash card
Requires : Software: Microsoft Windows 98 or newer, 4 MB RAM, 1 free USB port, CD drive
Description
Description
DicksonWare™ Software
  • Easy logger setup, data downloads and installation
  • For real-time monitoring and data visualization with tables and graphs
  • Can reset logger to factory settings
Card reader kit and Memory card
  • Transfers data from compatible loggers
  • Can be used where data logger cannot be brought back to computer
  • For transfering data from card to the DicksonWare software
Specifications
Specifications
Card reader and removable SD flash card
Card Reader Kit
Display Temperature and humidity logger, pressure datalogger
Documents