missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichlorotris(triphenylphosphine)ruthenium(II), 97%

$55.97 - $937.61

Chemical Identifiers

CAS 15529-49-4
Molecular Formula C54H45Cl2P3Ru
Molecular Weight (g/mol) 958.845
MDL Number MFCD00013077
InChI Key WIWBLJMBLGWSIN-UHFFFAOYSA-L
Synonym dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, tris triphenylphosphine ruthenium ii dichloride, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
PubChem CID 11007548
IUPAC Name dichlororuthenium;triphenylphosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Ru]Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0037303 View Documents Alfa Aesar™
L0037303*
1g Each for $55.97
 
AAL0037306 View Documents Alfa Aesar™
L0037306
5g Each for $220.53
 
AAL0037314 View Documents Alfa Aesar™
L0037314
25g Each for $937.61
 
Description

Description

Applications
Dichlorotris(triphenylphosphine)ruthenium(II) is used as a catalyst employed in a synthesis of furans from allenyl sulfides. C-H Activation Catalyst in a Ruthenium/Lewis Acid System. It is used in the Kharasch addition of chlorocarbons to alkenes.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

15529-49-4
958.845
WIWBLJMBLGWSIN-UHFFFAOYSA-L
11007548
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Ru]Cl
C54H45Cl2P3Ru
MFCD00013077
dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, tris triphenylphosphine ruthenium ii dichloride, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
dichlororuthenium;triphenylphosphane

Specifications

132°C to 134°C
15529-49-4
MFCD00013077
WIWBLJMBLGWSIN-UHFFFAOYSA-L
dichlororuthenium;triphenylphosphane
11007548
97%
1g
C54H45Cl2P3Ru
dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, tris triphenylphosphine ruthenium ii dichloride, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Ru]Cl
958.845
958.86
Dichlorotris(triphenylphosphine)ruthenium(II)