missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorotris(triphenylphosphine)ruthenium(II), 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0037314

 View more versions of this product

Catalog No. AAL0037314



Description

Description

Dichlorotris(triphenylphosphine)ruthenium(II) is used as a catalyst employed in a synthesis of furans from allenyl sulfides. C-H Activation Catalyst in a Ruthenium/Lewis Acid System. It is used in the Kharasch addition of chlorocarbons to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorotris(triphenylphosphine)ruthenium(II) is used as a catalyst employed in a synthesis of furans from allenyl sulfides. C-H Activation Catalyst in a Ruthenium/Lewis Acid System. It is used in the Kharasch addition of chlorocarbons to alkenes.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Dichlorotris(triphenylphosphine)ruthenium(II)
132°C to 134°C
MFCD00013077
Air and moisture sensitive
Insoluble in water.
[Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
958.85
958.86
15529-49-4
C54H45Cl2P3Ru
25 g
dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
KSHWMFNJCNGTJO-UHFFFAOYSA-L
dichlororuthenium;triphenylphosphane
11007548
97%

Chemical Identifiers

15529-49-4
958.85
KSHWMFNJCNGTJO-UHFFFAOYSA-L
11007548
C54H45Cl2P3Ru
MFCD00013077
dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
[Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 239-569-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only