Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

$210.88 - $635.72

Chemical Identifiers

Chemical Name or Material Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
CAS 92220-65-0
Molecular Formula C44H32Cl2Ir2N4
Formula Weight 1072.09
MDL Number MFCD07784515
Synonym [(ppy)2IrCl]2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27752MC

 
alfa aesar™
H27752MC
100mg Each for $210.88

AAH27752ME

 
alfa aesar™
H27752ME
500mg Each for $635.72
Description & Specifications

Specifications

Formula Weight 1072.09
Chemical Name or Material Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
Molecular Formula C44H32Cl2Ir2N4
MDL Number MFCD07784515
CAS 92220-65-0
Synonym [(ppy)2IrCl]2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.