missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
μ-Dichlorotetraethylene dirhodium(I), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197591000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
µ-Dichlorotetraethylene dirhodium(I) | |
Rust | |
Glass bottle | |
MFCD00013206 | |
chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i | |
OOQJCPOXJFCGCR-UHFFFAOYSA-L | |
ethene;rhodium;dichloride | |
45357812 | |
Crystalline Powder and/or Chunks |
12081-16-2 | |
Authentic | |
C8H16Cl2Rh2 | |
100 mg | |
Solubility in water: insoluble | |
Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C | |
388.93 | |
388.92 |
Chemical Identifiers
12081-16-2 | |
388.93 | |
OOQJCPOXJFCGCR-UHFFFAOYSA-L | |
45357812 | |
Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C |
C8H16Cl2Rh2 | |
MFCD00013206 | |
chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i | |
ethene;rhodium;dichloride |
Safety and Handling
AIR SENSITIVE
EINECSNumber : 235-145-0
RUO – Research Use Only