Learn More
Dichlorophenylsilane, 97%, AcroSeal™, Thermo Scientific Chemicals
$85.26 - $393.89
Chemical Identifiers
CAS | 80-10-4 |
---|---|
Molecular Formula | C12H10Cl2Si |
Molecular Weight (g/mol) | 253.20 |
MDL Number | MFCD00000489 |
InChI Key | OSXYHAQZDCICNX-UHFFFAOYSA-N |
Synonym | diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis |
PubChem CID | 6627 |
IUPAC Name | dichloro(diphenyl)silane |
SMILES | Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC430891000
|
Thermo Scientific Chemicals
430891000 |
100 mL | AcroSeal™ Glass Bottle |
N/A
|
|
||||
AC430898000
|
Thermo Scientific Chemicals
430898000 |
800 mL | AcroSeal™ Glass Bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
80-10-4 | |
253.20 | |
OSXYHAQZDCICNX-UHFFFAOYSA-N | |
6627 | |
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 |
C12H10Cl2Si | |
MFCD00000489 | |
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis | |
dichloro(diphenyl)silane |
Specifications
80-10-4 | |
Colorless to Yellow | |
305.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.5765 to 1.5815 | |
MFCD00000489 | |
1.204 | |
Solubility in water: decomposes | |
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 | |
253.20 | |
253.2 | |
Liquid |
-22.0°C | |
1.2040g/mL | |
155°C | |
96% min. (GC) | |
C12H10Cl2Si | |
(C6H5)2SiCl2 | |
100 mL | |
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis | |
OSXYHAQZDCICNX-UHFFFAOYSA-N | |
dichloro(diphenyl)silane | |
6627 | |
97% | |
Dichlorophenylsilane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
EINECSNumber : 201-251-
RUO – Research Use Only