missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorophenylsilane, 97%, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC430898000

 View more versions of this product

Catalog No. AC430898000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Dichlorophenylsilane
Clear colorless to yellow
80-10-4
AcroSeal™ Glass Bottle
Liquid
97%
(C6H5)2SiCl2
MFCD00000489
1.2040g/mL
800mL
Authentic
1.5765 to 1.5815 (20°C, 589nm)
≥96.0%
C12H10Cl2Si
253.2
Diphenyldichlorosilane

Chemical Identifiers

97%
(C6H5)2SiCl2
MFCD00000489
Dichlorophenylsilane
C12H10Cl2Si
253.2
Diphenyldichlorosilane
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store in a dry place. Store in a closed container. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

Danger