Please login to your online account to display your discounted pricing

Dichlorophenylphosphine 97%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  113665000

 View more versions of this product

Catalog No. AC113665000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 178.99
Physical Form Liquid
Quantity 500mL
Color Clear colorless to slightly yellow
EINECS Number 211-425-8
TSCA TSCA
RTECS Number TB2478000
Density 1.3190g/mL
Beilstein 16, 763
Infrared Spectrum Authentic
Synonym Phenylphosphonous dichloride
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Catches fire spontaneously if exposed to air.
Contact with water liberates toxic gas.
Reacts violently with water.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5970 to 1.5990 (20°C, 589nm)
Chemical Name or Material Dichlorophenylphosphine
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6 H5 Cl2 P
Linear Formula C6H5PCl2
MDL Number MFCD00000528
CAS 644-97-3

Specifications

Chemical Name or Material Dichlorophenylphosphine
Name Note 97%
CAS 644-97-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6 H5 Cl2 P
Linear Formula C6H5PCl2
Formula Weight 178.99
MDL Number MFCD00000528
Synonym Phenylphosphonous dichloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Catches fire spontaneously if exposed to air.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents