missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorophenylborane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 318550050

 View more versions of this product

Catalog No. AC318550050


 

Specifications Description & Specifications

Specifications

Dichlorophenylborane
Clear colorless to light colored
873-51-8
Glass bottle
Product darkens on storage
≥96.0% (ex Cl)
C6H5BCl2
158.82
1.2200g/mL
5g
Authentic
1.5450 to 1.5470 (20°C, 589nm)
Liquid
97%
C6H5BCl2
MFCD00013612

Chemical Identifiers

97%
C6H5BCl2
MFCD00013612
≥96.0% (ex Cl)
C6H5BCl2
158.82
Dichlorophenylborane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger