Please login to your online account to display your discounted pricing

Dichloromethylvinylsilane 97%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  147435000

 View more versions of this product

Catalog No. AC147435000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 141.07
Quantity 500g
Color Clear colorless to light brown
TSCA TSCA
RTECS Number VV3533000
Density 1.0800g/mL
EINECS Number 204-710-3
Synonym Methylvinyldichlorosilane
Refractive Index 1.4280 to 1.4310 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Health Hazard 1 Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD00000492
Chemical Name or Material Dichloromethylvinylsilane
Name Note 97%
Linear Formula H2C=CHSi(CH3)Cl2
Molecular Formula C3H6Cl2Si
CAS 124-70-9

Specifications

Chemical Name or Material Dichloromethylvinylsilane
Name Note 97%
CAS 124-70-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3H6Cl2Si
Linear Formula H2C=CHSi(CH3)Cl2
Formula Weight 141.07
MDL Number MFCD00000492
Synonym Methylvinyldichlorosilane
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents