Please login to your online account to display your discounted pricing

Dichloromethylvinylsilane 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  147430100

 View more versions of this product

Catalog No. AC147430100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Dichloromethylvinylsilane
Density 1.0800g/mL
Color Clear colorless to light brown
Quantity 10g
Name Note 97%
CAS 124-70-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number VV3533000
EINECS Number 204-710-3
TSCA TSCA
Infrared Spectrum Authentic
Physical Form Liquid
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3H6Cl2Si
Linear Formula H2C=CHSi(CH3)Cl2
Formula Weight 141.07
MDL Number MFCD00000492
Packaging Glass bottle
Refractive Index 1.4280 to 1.4310 (20°C, 589nm)
Synonym Methylvinyldichlorosilane

Specifications

Chemical Name or Material Dichloromethylvinylsilane
Name Note 97%
CAS 124-70-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3H6Cl2Si
Linear Formula H2C=CHSi(CH3)Cl2
Formula Weight 141.07
MDL Number MFCD00000492
Synonym Methylvinyldichlorosilane
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents