Please login to your online account to display your discounted pricing

Dichloromethylsilane, 97%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  147395000

 View more versions of this product

Catalog No. AC147395000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 115.04
Physical Form Liquid
Quantity 500mL
Color Clear light yellow to light brown
Beilstein 04, I, 581
Density 1.1000g/mL
TSCA TSCA
RTECS Number VV3500000
EINECS Number 200-877-1
Refractive Index 1.3970 to 1.4000 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym Methyldichlorosilane
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Fatal if inhaled.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently with water.
Linear Formula CH3SiHCl2
CAS 75-54-7
Name Note 97%
MDL Number MFCD00000494
Chemical Name or Material Dichloromethylsilane
Molecular Formula CH4Cl2Si
Assay Percent Range 97%

Specifications

Chemical Name or Material Dichloromethylsilane
Name Note 97%
CAS 75-54-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula CH4Cl2Si
Linear Formula CH3SiHCl2
Formula Weight 115.04
MDL Number MFCD00000494
Synonym Methyldichlorosilane
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Fatal if inhaled.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents