missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethylsilane, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01407936

 View more versions of this product

Catalog No. AA1407936


Description

Description

A monomer (‘building block’) in the production of silicone polymers. An intermediate (starting material) in the production of other organic and inorganic chemicals. In the electronics industry for the production of ultra-pure polysilicon and in the manufacture of semiconductors and photovoltaics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

41°C to 42°C
-93°C
Colorless
500 g
MFCD00000494
1071194
Methyldichlorosilane
115.03
1.398
1.105 g/mL
Dichloromethylsilane
75-54-7
Liquid
CH3SiHCl2
UN1242
-32°C (-26°F)
Soluble in heptane,benzene,ether.
≥97%
Air and moisture sensitive

Chemical Identifiers

75-54-7
MFCD00000494
Methyldichlorosilane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H260-H314-H318-H332
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P210-P223-P231+P232-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H260-H301-H314-H330

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METHYLDICHLOROSILANE

EINECSNumber : 200-877-1

RTECSNumber : VV3500000

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only