Please login to your online account to display your discounted pricing

Dichloromethylphenylsilane 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dichloromethylphenylsilane
Name Note 98%
CAS 149-74-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8Cl2Si
Linear Formula C6H5Si(CH3)Cl2
Formula Weight 191.13
MDL Number MFCD00000490
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC147385000

  SDS
thermo fisher scientific
147385000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC147380100

  SDS
thermo fisher scientific
147380100
Glass bottle 10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC147381000

  SDS
thermo fisher scientific
147381000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 191.13
Quantity 500mL
Color Clear colorless
EINECS Number 205-746-2
TSCA TSCA
RTECS Number VV3530000
Density 1.1760g/mL
Packaging Glass bottle
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Dichloromethylphenylsilane
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H8Cl2Si
Linear Formula C6H5Si(CH3)Cl2
MDL Number MFCD00000490
CAS 149-74-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.