missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethylphenylsilane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2399914

 View more versions of this product

Catalog No. AAB2399914



Description

Description

Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is sensitive to moisture. Protect from humidity and water.
Specifications

Specifications

205°C to 206°C
149-74-6
C7H8Cl2Si
MFCD00000490
970975
methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
GNEPOXWQWFSSOU-UHFFFAOYSA-N
dichloro-methyl-phenylsilane
9006
98%
1.519
1.173
Dichloromethylphenylsilane
25 g
C6H5Si(CH3)Cl2
UN2437
82°C (179°F)
Reacts with water.
C[Si](Cl)(Cl)C1=CC=CC=C1
191.13
191.13
Acrid
Moisture sensitive

Chemical Identifiers

149-74-6
191.13
GNEPOXWQWFSSOU-UHFFFAOYSA-N
9006
C[Si](Cl)(Cl)C1=CC=CC=C1
C7H8Cl2Si
MFCD00000490
methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
dichloro-methyl-phenylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P270-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHYLPHENYLDICHLOROSILANE

EINECSNumber : 205-746-2

RTECSNumber : VV3530000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only