Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloromethyl-n-octylsilane, 98%

Chemical Identifiers

Chemical Name or Material Dichloromethyl-n-octylsilane
CAS 14799-93-0
Molecular Formula C9H20Cl2Si
Formula Weight 227.25
MDL Number MFCD00042606
UN Number UN2987
Beilstein 1740697
Synonym n-Octylmethyldichlorosilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1656509

  SDS
alfa aesar™
L1656509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1656518

  SDS
alfa aesar™
L1656518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.25
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 98%
Chemical Name or Material Dichloromethyl-n-octylsilane
Beilstein 1740697
Quantity 10g
MDL Number MFCD00042606
Molecular Formula C9H20Cl2Si
CAS 14799-93-0
Boiling Point 224° to 226°C
Synonym n-Octylmethyldichlorosilane
TSCA TSCA
UN Number UN2987
Health Hazard 1 Danger
Flash Point 98°C (208°F)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.