Please login to your online account to display your discounted pricing

Alfa Aesar™ (Dichloromethyl)dimethylchlorosilane, 94%

Chemical Identifiers

Chemical Name or Material (Dichloromethyl)dimethylchlorosilane
CAS 18171-59-0
Molecular Formula C3H7Cl3Si
Linear Formula Cl2CHSi(CH3)2Cl
Formula Weight 177.53
MDL Number MFCD00018852
UN Number UN2986
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2222509

  SDS
alfa aesar™
B2222509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2222518

  SDS
alfa aesar™
B2222518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.53
UN Number UN2986
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Assay 94%
Health Hazard 1 Danger
Molecular Formula C3H7Cl3Si
Linear Formula Cl2CHSi(CH3)2Cl
Chemical Name or Material (Dichloromethyl)dimethylchlorosilane
CAS 18171-59-0
Flash Point 27°C (81°F)
Boiling Point 148° to 150°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
TSCA TSCA
Quantity 10g
MDL Number MFCD00018852
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.