Please login to your online account to display your discounted pricing

Dichloromethane UltimAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Dichloromethane
CAS 75-09-2
Molecular Formula CH2Cl2
Formula Weight 84.93
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MKH4851

 
avantor performance
H48506
Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MKH4854

 
avantor performance
H48510
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dichloromethane
CAS 75-09-2
Molecular Formula CH2Cl2
Formula Weight 84.93
Packaging Glass
Quantity 1L