Please login to your online account to display your discounted pricing

Dichloromethane UltimAR, Macron Fine Chemicals™


macron

Quantity 1L
Packaging Glass

Manufacturer: avantor performance  H48506

 View more versions of this product

Catalog No. MKH4851

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 84.93
Chemical Name or Material Dichloromethane
Packaging Glass
Quantity 1L
Molecular Formula CH2Cl2
CAS 75-09-2

Specifications

Chemical Name or Material Dichloromethane
CAS 75-09-2
Molecular Formula CH2Cl2
Formula Weight 84.93