missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene

Manufacturer:  Alfa Aesar™ 32440M1

 View more versions of this product

Catalog No. AA32440M1



Specifications

Specifications

Dichloromethane
75-09-2
MFCD00000881
1730800
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
C(Cl)Cl
84.927
CHEBI:15767
Liquid
Sprectrophotometric
Sweet
39°C to 40°C
4 x 1L
Stabilized with amylene
CH2Cl2
UN1593
14,6063
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
6344
84.93
≥99.7%
1.325g/mL
1.4244
-95°C

Chemical Identifiers

75-09-2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
dichloromethane
CH2Cl2
MFCD00000881
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
CHEBI:15767
C(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE

RTECSNumber : PA8050000

TSCA : TSCA

SDS Documents