Please login to your online account to display your discounted pricing

Dichloromethane For spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$88.34 - $332.40

Chemical Identifiers

Chemical Name or Material Dichloromethane
CAS 75-09-2
Molecular Formula CH2Cl2
Formula Weight 84.93
UN Number 1593
Synonym DCM, Methylene dichloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1060480500

 
emd millipore corporation
1060480500
Glass Bottle 500mL Each for $88.34

M1060482500

 
emd millipore corporation
1060482500
Glass Bottle 2.5L Each for $332.40
Description & Specifications

Specifications

Formula Weight 84.93
Boiling Point 40°C
Melting Point −95°C
pH Neutral (20°C)
Quantity 500mL
Vapor Pressure 475 hPa (20°C)
Molecular Formula CH2Cl2
Synonym DCM, Methylene dichloride
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Packaging Glass Bottle
Odor Sweet
Chemical Name or Material Dichloromethane
Density 1.33g/cm3 at 20°C
Storage Information 5°C to 30°C (41°F to 86°F), Tightly closed. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons. Protected from light.
Health Hazard 1 H351: Suspected of causing cancer.
Solubility Information Soluble in water: 20g/L at 20°C
Refractive Index 1.42
Physical Form Liquid
Ignition Point 605°C (1121°F)
Color Colorless
CAS 75-09-2
UN Number 1593
Safety and Handling

H351: Suspected of causing cancer.