Please login to your online account to display your discounted pricing

Dichloromethane-d2, for NMR, packaged in 1.00 ml ampoules, 99.6+ atom % D, ACROS Organics™

$340.54 - $340.54

Chemical Identifiers

Chemical Name or Material Dichloromethane-d2
Name Note 99.6+ atom % D
CAS 1665-00-5
Purity Grade For NMR
Molecular Formula CCl2D2
Linear Formula CD2Cl2
Formula Weight 86.95
MDL Number MFCD00000882
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC217350100

  SDS
thermo fisher scientific
217350100
Ampule 10 x 1.00mL Each for $340.54
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 86.95
Quantity 10 x 1.00mL
Color Clear colorless
EINECS Number 216-776-0
Density 1.3600g/mL
Purity Grade For NMR
Beilstein 01, IV, 39
Water 0.04% max. (Coulometric)
Chemical Name or Material Dichloromethane-d2
Name Note 99.6+ atom % D
Molecular Formula CCl2D2
Linear Formula CD2Cl2
MDL Number MFCD00000882
CAS 1665-00-5
Deuterium (D) 99.60% min.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Ampule

For NMR

Safety and Handling

Warning

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.