missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethane-d2, for NMR, 99.5 atom % D, AcroSeal™, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 433991000

Catalog No. AC433991000


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
Specifications

Specifications

Dichloromethane-d2
1665-00-5
Colorless
40.0°C
CCl2D2
CD2Cl2
1.36
Solubility in water: .. Other solubilities: soluble in alchol,ether and dmf
C(Cl)Cl
86.95
86.95
Liquid
For NMR
-97.0°C
1.3600g/mL
AcroSeal™ Glass Bottle
1.4218
100 mL
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
YMWUJEATGCHHMB-DICFDUPASA-N
dichloro(dideuterio)methane
160586
NMR

Chemical Identifiers

1665-00-5
86.95
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
dichloro(dideuterio)methane
CCl2D2
YMWUJEATGCHHMB-DICFDUPASA-N
160586
C(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 216-776-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only