Please login to your online account to display your discounted pricing

Dichloromethane - D2, deuteration degree min. 99.96% for NMR spectroscopy MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Packaging Glass ampoule

Manufacturer: emd millipore corporation  1.04200.0009

 View more versions of this product

Catalog No. M1042000009

  • / Each
 

Description & Specifications

Chemical Identifiers

Odor Sweet
Packaging Glass ampoule
Ignition Point 605°C (1121°F)
Vapor Pressure ca. 450 hPa at 20°C (68°F)
Quantity 10 x 0.75mL
Color Colorless
Chemical Name or Material Dichloromethane - D2
CAS 1665-00-5
Physical Form Liquid
Assay Percent Range 99.96% min.
Molecular Formula CD2Cl2
Formula Weight 86.95g/mol
Synonym Dideuteromethylene chloride

Specifications

Chemical Name or Material Dichloromethane - D2
CAS 1665-00-5
Physical Form Liquid
Assay Percent Range 99.96% min.
Molecular Formula CD2Cl2
Formula Weight 86.95g/mol
Synonym Dideuteromethylene chloride