Please login to your online account to display your discounted pricing

Dichloromethane - D2, deuteration degree min. 99.96% for NMR spectroscopy MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$654.84 - $654.84

Chemical Identifiers

Chemical Name or Material Dichloromethane - D2
CAS 1665-00-5
Physical Form Liquid
Assay Percent Range 99.96% min.
Molecular Formula CD2Cl2
Formula Weight 86.95
Synonym Dideuteromethylene chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1042000010

 
emd millipore corporation
1042000010
Glass ampoule 10mL Each for $654.84
Description & Specifications

Specifications

Chemical Name or Material Dichloromethane - D2
CAS 1665-00-5
Odor Sweet
Packaging Glass ampoule
Ignition Point 605°C (1121°F)
Vapor Pressure ca. 450 hPa at 20°C (68°F)
Physical Form Liquid
Quantity 10mL
Assay Percent Range 99.96% min.
Color Colorless
Molecular Formula CD2Cl2
Formula Weight 86.95
Synonym Dideuteromethylene chloride