Please login to your online account to display your discounted pricing

Dichloromethane - D2, deuteration degree min. 99.96% for NMR spectroscopy MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 10 x 0.5mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  1042000005

 View more versions of this product

Catalog No. M1042000005

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 86.95
Quantity 10 x 0.5mL
Color Colorless
Physical Form Liquid
Packaging Glass ampoule
Assay Percent Range 99.96% min.
Vapor Pressure ca. 450 hPa at 20°C (68°F)
Odor Sweet
Molecular Formula CD2Cl2
Synonym Dideuteromethylene chloride
Ignition Point 605°C (1121°F)
Chemical Name or Material Dichloromethane - D2
CAS 1665-00-5

Specifications

Chemical Name or Material Dichloromethane - D2
CAS 1665-00-5
Physical Form Liquid
Assay Percent Range 99.96% min.
Molecular Formula CD2Cl2
Formula Weight 86.95
Synonym Dideuteromethylene chloride