Please login to your online account to display your discounted pricing

Dichloromethane-D2, deuteration degree min. 99.8% for NMR spectroscopy MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Packaging Glass ampoule

Manufacturer: emd millipore corporation  1.13720.0010

 View more versions of this product

Catalog No. M1137200010

  • / Each
 

Description & Specifications

Chemical Identifiers

Odor Sweet
Packaging Glass ampoule
Vapor Pressure 450 hPa at 20°C (68°F)
Quantity 10mL
Color Colorless
Chemical Name or Material Dichloromethane-D2
CAS 1665-00-5
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CD2Cl2
Formula Weight 86.95g/mol

Specifications

Chemical Name or Material Dichloromethane-D2
CAS 1665-00-5
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CD2Cl2
Formula Weight 86.95g/mol