Please login to your online account to display your discounted pricing

Dichloromethane-D2, deuteration degree min. 99.8% for NMR spectroscopy MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 10 x 0.75mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  1137200009

 View more versions of this product

Catalog No. M1137200009

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 86.95
Quantity 10 x 0.75mL
Color Colorless
Physical Form Liquid
Vapor Pressure 450 hPa at 20°C (68°F)
Molecular Formula CD2Cl2
Packaging Glass ampoule
Assay Percent Range 99.8% min.
Odor Sweet
Chemical Name or Material Dichloromethane-D2
CAS 1665-00-5

Specifications

Chemical Name or Material Dichloromethane-D2
CAS 1665-00-5
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CD2Cl2
Formula Weight 86.95