missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethane-D2, deuteration degree min. 99.8% for NMR spectroscopy MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$328.81 - $328.81

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.939
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
PubChem CID 160586
IUPAC Name dichloro(dideuterio)methane
SMILES C(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1137200009 View Documents Millipore Sigma
1137200009
10 x 0.75mL Glass ampoule Each for $328.81
Specifications

Chemical Identifiers

1665-00-5
86.939
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
dichloro(dideuterio)methane
CH2Cl2
YMWUJEATGCHHMB-DICFDUPASA-N
160586
C(Cl)Cl

Specifications

Sweet
450 hPa at 20°C (68°F)
10 x 0.75mL
99.8% min.
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
C(Cl)Cl
86.939
86.95
Dichloromethane-D2
Glass ampoule
Colorless
1665-00-5
CH2Cl2
YMWUJEATGCHHMB-DICFDUPASA-N
dichloro(dideuterio)methane
160586
Liquid