Learn More
Dichloromethane, ACS, 99.5+%, stab. with amylene, Thermo Scientific Chemicals
Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, Free halogens P.T., H2O 0.02%
Supplier: Thermo Scientific Chemicals 039116K7
Description
Dichloromethane is a chlorinated hydrocarbon that has been employed as a pharamacuetical applications. Mainly, it's utilization as a solvent in manufacturing and food engineering. It is too useful in the drinking bird heat engine. It was the solvent of choice for caffeine extraction. It is likewise employed as a solvent and paint remover. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dichloromethane | |
Colorless | |
1.325 g/cm3 at 20°C | |
4 L | |
Sweet | |
1.4244 | |
MFCD00000881 | |
1730800 | |
Miscible in ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. | |
ClCCl | |
84.93 | |
ACS |
75-09-2 | |
Colorless liquid | |
39°C to 40°C | |
605°C (1121°F) | |
Liquid | |
CH2Cl2 | |
475 hPa (356mm Hg) at 20°C | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
84.93 |
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
ClCCl |
Safety and Handling
P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H336-H350-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 200-838-9
RTECSNumber : PA8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only