Please login to your online account to display your discounted pricing

Dichloromethane 99+%, stabilized with amylene, pure, ACROS Organics™


Quantity 5L

Manufacturer: thermo fisher scientific  AC383780050

 View more versions of this product

Catalog No. AC383780050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point −97°C
Formula Weight 84.93
Quantity 5L
Packaging Glass Bottle
Density 1.325g/cm3
EINECS Number 200-838-9
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
MDL Number MFCD00000881
CAS 75-09-2
Chemical Name or Material Dichloromethane
Molecular Formula CH2Cl2
Assay Percent Range 99+%
Synonym Methylene chloride

Specifications

Chemical Name or Material Dichloromethane
Assay Percent Range 99+%
Molecular Formula CH2Cl2
Formula Weight 84.93
MDL Number MFCD00000881
Synonym Methylene chloride