Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloromethane, 99+%, stab. with ca. 50ppm amylene

$39.39 - $90.81

Chemical Identifiers

Chemical Name or Material Dichloromethane
CAS 75-09-2
Name Note stabilized with ca. 50ppm amylene
Molecular Formula CH2Cl2
Formula Weight 84.93
MDL Number MFCD00000881
UN Number UN1593
Beilstein 1730800
Merck Index 14,6063
Synonym DCM; Methylene chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL130890F

 
alfa aesar™
L130890F
2500mL Each for $90.81

AAL13089AP

 
alfa aesar™
L13089AP
500mL Each for $39.39
Description & Specifications

Specifications

Formula Weight 84.93
Chemical Name or Material Dichloromethane
Name Note stabilized with ca. 50ppm amylene
Boiling Point 39° to 40°C
Quantity 2500mL
Assay ≥99%
Molecular Formula CH2Cl2
MDL Number MFCD00000881
CAS 75-09-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Odor Sweet
Melting Point −95°C
UN Number UN1593
Beilstein 1730800
Merck Index 14,6063
Synonym DCM; Methylene chloride
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE
RTECS Number PA8050000

Applications
Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

Solubility
Miscible with ethyl acetate, alcohol, hexanes, methanol, diethyl ether, n-octanol, acetone benzene, carbon tetrachloride, diethyl ether and chloroform. Immiscible with water.

Notes
Keep the container tightly closed in a cool, well-ventilated area. Incompatible with alkali metals, aluminum, strong oxidizing agents, bases, amines, magnesium, strong acids strong bases and vinyl compounds.

Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.