Learn More
Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, Ac, Thermo Scientific Chemicals
Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, AcroSeal™.
Supplier: Thermo Scientific Chemicals 406910025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dichloromethane | |
75-09-2 | |
39.0°C to 40.0°C | |
AcroSeal™ Glass Bottle | |
1.325 | |
CH2Cl2 | |
0.43 mPa.s (20°C) | |
01,676; 02,273; 04,337; 07,239; 13,106 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
84.93 | |
Biochemical |
-97.0°C | |
1.3250g/mL | |
2.5 L | |
99.85% min. (GC) | |
CH2Cl2 | |
MFCD00000881 | |
01,6 | |
15,6135 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and dmf | |
ClCCl | |
84.93 | |
99.9% |
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
ClCCl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
May cause drowsiness or dizziness.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
EINECSNumber : 200-838-9
RUO – Research Use Only