Please login to your online account to display your discounted pricing

Dichloromethane, 99.8% min., EMD Millipore

$102.63 - $102.63

Chemical Identifiers

Chemical Name or Material Dichloromethane
CAS 75-09-2
Name Note For HPLC, ACS analysis
Molecular Formula CH2Cl2
Formula Weight 84.93
Synonym Methylene Chloride
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

MDX08386

 
emd millipore corporation
DX08386
Each for $102.63
Description & Specifications

Specifications

Chemical Name or Material Dichloromethane
Name Note For HPLC, ACS analysis
CAS 75-09-2
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CH2Cl2
Formula Weight 84.93
Synonym Methylene Chloride
Color Clear colorless
Quantity 1L